missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) selenide, 99.999+% (metals basis)

Catalog No. AA4491622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4491622 100 g
AA4491614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4491622 Supplier Thermo Scientific Chemicals Supplier No. 04491622
Only null left

CAS: 12069-00-0 | PbSe | 286.20 g/mol

Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12069-00-0
Molecular Formula PbSe
Molecular Weight (g/mol) 286.20
MDL Number MFCD00016273
InChI Key GGYFMLJDMAMTAB-UHFFFAOYSA-N
Synonym lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
PubChem CID 61550
IUPAC Name selanylidenelead
SMILES [Se]=[Pb]

Specifications

Color Gray
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range >99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 286.15
Chemical Name or Material Lead(II) selenide
Hazard Category H301+H331-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 235-109-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only