missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) selenide, 99.999+% (metals basis)
Description
Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Physical Form | Fused Lump |
| Quantity | 25 g |
| Assay Percent Range | >99.999% (Metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 286.15 |
| Chemical Name or Material | Lead(II) selenide |
Safety and Handling
| Hazard Category | H301+H331-H350-H360Df-H373 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 235-109-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only