missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Phthalocyanine 95.0+%, TCI America™
Specifications
Specifications
| Color | Blue-Purple |
| Quantity | 1 g |
| UN Number | 3077 |
| Formula Weight | 719.74 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lead(II) Phthalocyanine |
Safety and Handling
| TSCA | Yes |