missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) perchlorate trihydrate, ACS reagent

Catalog No. AC447785000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447785000 500 g Plastic bottle
AC447781000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447785000 Supplier Thermo Scientific Chemicals Supplier No. 447785000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.10
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name lead(2+);diperchlorate;trihydrate
SMILES O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 100.0°C
Quantity 500 g
Linear Formula Pb(ClO4)2·3H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 460.14
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.6
Density 2.6000g/mL
Chemical Name or Material Lead(II) perchlorate trihydrate
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only