missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%

Catalog No. p-7048842
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1164022 100 g
AA1164036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1164022 Supplier Thermo Scientific Chemicals Supplier No. 01164022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Titrant in complexometric titrations | CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.10
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name lead(2+);diperchlorate;trihydrate
SMILES O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 100°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Pb(ClO4)2·3H2O
UN Number UN1470
Solubility Information Soluble in water and alcohol.
Formula Weight 460.15 (406.10 Anhydrous)
Percent Purity ≤97%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 2.60 g/mL
Chemical Name or Material Lead(II) perchlorate trihydrate
Show More Show Less
Hazard Category H272-H302+H332-H336-H350-H360Df-H373
Hazard Statement GHS H Statement
H271-H360-H373-H302-H332
May cause fire or explosion
strong oxidizer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LEAD PERCHLORATE, SOLID
EINECSNumber 237-125-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only