missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Honeywell Fluka™

Puriss. p.a.,≥99.0% (KT), yellow

$543.24 - $543.24

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044873
View Documents
Honeywell Chemicals
153381KG
1 kg Poly Bottle
Each for $543.24
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1317-36-8
223.20
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
O=[Pb]

Specifications

1470°C
1317-36-8
Solid
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
O=[Pb]
223.20
223.2g/mol
puriss. p.a.
10mmHg (0°C)
Lead(Ii) Oxide
890°C
Yellow
1 kg
PbO
UN2291
YEXPOXQUZXUXJW-UHFFFAOYSA-N
plumbanone
14827
≥99.0% (KT)
Poly Bottle
9.66g/cu.cm
Videos
Safety and Handling

Safety and Handling

P201-P260-P263-P280-P301 + P312 + P330-P308 + P313

H302 + H332-H351-H360Df-H362-H372-H410

DOTInformation : 6.1/PGIII

EINECSNumber : 215-267-0

RTECSNumber : OG1750000

SDS
missing translation for 'documents'

missing translation for 'documents'