missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Puratronic™, 99.999% (metals basis)

Catalog No. p-7051209
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4075314 25 g
AA4075322 100 g
AA4075336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4075314 Supplier Thermo Scientific Chemicals Supplier No. 04075314
Only null left

CAS: 1317-36-8 | OPb | 223.20 g/mol

Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]

Specifications

Melting Point 886°C
Color Yellow
Physical Form Powder
Quantity 25 g
UN Number UN3077
Merck Index 14,5413
Solubility Information Insoluble in water,soluble in concentrated alkalies,hydrochloric acid and ammonium chloride.
Formula Weight 223.19
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 9.53 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) oxide
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H372
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-267-0
RTECSNumber OG1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only