missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Oxide, Extra Pure, MilliporeSigma™

Catalog No. p-4939048
Change view
Click to view available options
Quantity:
5 kg
50 kg
Packaging:
Plastic bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1056585000 5 kg Plastic bottle
M1056589050 50 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1056585000 Supplier Technidata Supplier No. 1.05658.5000
Only null left

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name plumbanone
SMILES O=[Pb]

Specifications

Physical Form Solid
Quantity 5 kg
UN Number UN 2291
Formula Weight 223.19
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Appearance Yellow solid
Chemical Name or Material Lead(II) oxide
Hazard Category H360FD: May damage fertility. May damage the unborn child.
H302 + H332: Harmful if swallowed or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-267-0