missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder

CAS: 1317-36-8 | OPb | 223.20 g/mol

Supplier:  Thermo Scientific Chemicals 315850025

 View more versions of this product

Catalog No. AC315850025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis
Specifications

Specifications

1470.0°C
886.0°C
1317-36-8
Powder
99.9+%
PbO
01,536; 02,233
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
YEXPOXQUZXUXJW-UHFFFAOYSA-N
plumbanone
14827
≥99.9%
Glass bottle
Lead(II) oxide
<10 micron, 99.9+%
Yellow
2.5 kg
OPb
MFCD00011164
15, 5468
Solubility in water: 70.2mg/l (20°C)
O=[Pb]
223.20
223.19
Trace Metal Basis

Chemical Identifiers

1317-36-8
223.20
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
O=[Pb]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.

GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C

GHS Signal Word: Danger

EINECSNumber : 215-267-0

RTECSNumber : OG1750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only