missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder

Catalog No. AC315851000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315851000 100 g Glass bottle
AC315855000 500 g Glass bottle
AC315850025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315851000 Supplier Thermo Scientific Chemicals Supplier No. 315851000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1317-36-8 | OPb | 223.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Boiling Point 1470.0°C
Melting Point 886.0°C
Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9+%
Linear Formula PbO
Fieser 01,536; 02,233
Merck Index 15, 5468
Solubility Information Solubility in water: 70.2 mg/l (20°C).
IUPAC Name oxolead
Formula Weight 223.19
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lead(II) oxide, <10 micron, 99.9+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
EINECSNumber 215-267-0
RTECSNumber OG1750000
TSCA TSCA

RUO – Research Use Only