missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99.9%

CAS: 1317-36-8 | OPb | 223.20 g/mol

Supplier:  Thermo Scientific Chemicals 012220A3

 View more versions of this product

Catalog No. AA12220A3


Only null left
Explore available promotions

Description

Description

It is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. Lead monoxide is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) oxide
1317-36-8
Powder
99.9%
MFCD00011164
Soluble in concentrated alkali,hydrochloric acid,and ammonium chloride. Insoluble in water,dilute alkali and alcohol.
O=[Pb]
223.20
223.19
9.53
886°C
Yellow
2 kg
OPb
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
YEXPOXQUZXUXJW-UHFFFAOYSA-N
oxolead
14827
Odorless

Chemical Identifiers

1317-36-8
223.20
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
O=[Pb]
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
oxolead
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H372

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-267-0

RTECSNumber : OG1750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only