missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-4824786
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1072718 50 g
AA1072730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1072718 Supplier Thermo Scientific Chemicals Supplier No. 01072718
Only null left

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol

Employed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds application in supramolecular chemistry. Strong oxidizing agent, and an alternative to the Sommelet reaction for the oxidation of benzylic halides to aldehydes. Used in the preparation of isothiocyanates from dithiocarbamates. Scavenger of bromide in nucleophilic substitution reactions. Manufacture of dinitrogen tetroxide in indurtrial scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2
Odor Odorless
Sensitivity Hygroscopic
Density 4.53
Chemical Name or Material Lead(II) nitrate
Show More Show Less
Hazard Category H272-H302+H332-H318-H336-H350-H360Df-H373
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 233-245-9
RTECSNumber OG2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only