missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, 99+%, ACS reagent

Catalog No. AC423850050
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423850050 5 g Glass Bottle
AC423855000 500 g Plastic bottle
AC423850025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423850050 Supplier Thermo Scientific Chemicals Supplier No. 423850050

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula Pb(NO3)2
Merck Index 15, 5469
Solubility Information Solubility in water: 343g/L.
IUPAC Name λ²-lead(2+) dinitrate
Formula Weight 331.2
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Specific Gravity 4.53
Density 4.5300g/mL
Chemical Name or Material Lead(II) nitrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume
EINECSNumber 233-245-9
RTECSNumber OG2100000
TSCA TSCA

RUO – Research Use Only