missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, ACS, 99.0% min

Catalog No. p-4407544
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1424336 500 g
AA14243A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1424336 Supplier Thermo Scientific Chemicals Supplier No. 01424336

For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile | CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol

As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photographyEmployed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Freely soluble in water. Soluble in absolute alcohol,methanol. Insoluble in concentrated HNO3
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Vapor Pressure approx.3.0-4.0 (20% aqueous solution)
Density 4.53 g/cm3 at 20°C
Chemical Name or Material Lead(II) nitrate, For ACS analysis
Show More Show Less
Hazard Category H272-H302+H332-H318-H336-H350-H360Df-H373
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 233-245-9
RTECSNumber OG2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only