Learn More
Lead(II) metaborate monohydrate, Thermo Scientific Chemicals
$658.47 - $658.47
Description
Lead(II) metaborate monohydrate acts as a starting material used in the preparation of lead(II) borates such as Pb2B3O5.5 (OH)2 and [Pb3(B3O7)](NO3) by changing the pH values of the reaction systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10214-39-8 | |
| MFCD00049504 | |
| Lead(II) borate |
Specifications
| 160°C (in -H2O) | |
| Powder | |
| PbB2O4·H2O | |
| UN2291 | |
| Insoluble in water and alkali; Soluble in acids. | |
| Odorless | |
| Lead(II) metaborate monohydrate |
| 10214-39-8 | |
| 100 g | |
| MFCD00049504 | |
| Lead(II) borate | |
| 310.81 (292.81 Anhydrous) | |
| 5.598 g/mL |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber : 238-762-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only