missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) metaborate monohydrate, Thermo Scientific Chemicals

Catalog No. AA3933922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3933922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3933922 Supplier Thermo Scientific Chemicals Supplier No. 03933922

Lead(II) metaborate monohydrate acts as a starting material used in the preparation of lead(II) borates such as Pb2B3O5.5 (OH)2 and [Pb3(B3O7)](NO3) by changing the pH values of the reaction systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10214-39-8
MDL Number MFCD00049504
Synonym Lead(II) borate

Specifications

Melting Point 160°C (in -H2O)
Physical Form Powder
Quantity 100 g
Linear Formula PbB2O4·H2O
UN Number UN2291
Solubility Information Insoluble in water and alkali; Soluble in acids.
Formula Weight 310.81 (292.81 Anhydrous)
Odor Odorless
Density 5.598 g/mL
Chemical Name or Material Lead(II) metaborate monohydrate
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 238-762-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only