missing translation for 'onlineSavingsMsg'
Learn More

Lead(II,IV) oxide, red, 98%

Catalog No. p-44166
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221111000 100 g Glass bottle
AC221115000 500 g Glass bottle
AC221110051 5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221111000 Supplier Thermo Scientific Chemicals Supplier No. 221111000

CAS: 1314-41-6 | O4Pb3 | 685.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.57
MDL Number MFCD00078491
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
Synonym lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
PubChem CID 16685188
IUPAC Name 1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
SMILES O1[Pb]O[Pb]12O[Pb]O2

Specifications

Melting Point 500.0°C
Color Orange
Physical Form Powder
Quantity 100 g
Assay Percent Range 97.5% min. (PB3O4)
Linear Formula 2PbO·PbO2
Merck Index 15,548
Solubility Information Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol
Formula Weight 685.57
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Lead(II, IV) oxide, red, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and k
EINECSNumber 215-235-6
RTECSNumber OG5425000
TSCA TSCA

RUO – Research Use Only