missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Iodide (99.99%, trace metals basis) 98.0+%, TCI America™

Catalog No. p-7131908
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02791G 1 g
L02795G 5 g
L027925G 25 g
L0279100G 100 g
L02791KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. L02791G Supplier TCI America Supplier No. L02791G
Only null left

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Boiling Point 954°C
Melting Point 412°C
Physical Form Crystalline Powder
Quantity 1 g
UN Number 2291
Formula Weight 461.01
Percent Purity ≥98.0% (T)
Chemical Name or Material Lead(II) Iodide (99.99%, trace metals basis) [for Perovskite precursor]
EINECSNumber (1)-1139
TSCA Yes