missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) iodide, 98.5%

Catalog No. p-4407542
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA5710318 50 g
AA5710330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA5710318 Supplier Thermo Scientific Chemicals Supplier No. 05710318
Only null left

CAS: 10101-63-0 | I2Pb | 461.00 g/mol

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Boiling Point 954°C
Melting Point 402°C
Color Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 98.5%
Solubility Information Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides and sodium thiosulfate. Insoluble in alcohol,cold hydrochloric acid and organic solvents.
Formula Weight 461
Odor Odorless
Sensitivity Light sensitive
Appearance Yellow powder
Density 6.16 g/cm3 at 20°C
Chemical Name or Material Lead(II) iodide
Show More Show Less
Hazard Category H302+H332-H350-H360-H373
Precautionary Statement P201-P202-P261-P262-P264b-P270-P271-P280-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-256-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only