missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Carbonate Basic, Honeywell Fluka™

Puriss., white lead,≥77% Pb basis

$441.08 - $18596.80

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001907
View Documents
Honeywell Chemicals
115131KG
1 kg
Each for $441.08
Only null left
 
6000892
View Documents
Honeywell Chemicals
1151350KG
50 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1319-46-6
MFCD00078155

Specifications

1319-46-6
(PbCO3)2·Pb(OH)2
3077
≥77% (Pb basis)
Lead(Ii) Carbonate Basic
1 kg
MFCD00078155
775.63g/mol
puriss.
Videos
Safety and Handling

Safety and Handling

P201-P260-P280-P301 + P312 + P330-P308 + P313

H302 + H332-H360Df-H373-H410

EINECSNumber : 215-290-6

RTECSNumber : OF9275000

SDS
missing translation for 'documents'

missing translation for 'documents'