missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, basic, extra pure

Catalog No. p-3734839
Change view
Click to view available options
Quantity:
10 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Catalog No. Quantity Packaging
AC222710025 2.5 kg Plastic bottle
AC222710100 10 kg Plastic drum
2 options

Catalog No. AC222710025

Supplier: Thermo Scientific Chemicals 222710025

Only null left

CAS: 1319-46-6 | C2H2O8Pb3 | 775.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
Molecular Weight (g/mol) 775.59
MDL Number MFCD00078155

Specifications

Melting Point 400.0°C
Color White
Physical Form Powder
Quantity 2.5 kg
Linear Formula (PbCO3)2·Pb(OH)2
Merck Index 15, 1002
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 775.59
Percent Purity ≥78% (Pb)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lead(II) carbonate, basic, -325 Mesh
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b
EINECSNumber 215-290-6

RUO – Research Use Only