missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, basic, 99% (metals basis)

Catalog No. AA4208936
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4208936 500 g
AA42089A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4208936 Supplier Thermo Scientific Chemicals Supplier No. 04208936

CAS: 1319-46-6 | C2H2O8Pb3

Lead(II) carbonate is employed in the production of anticorrosion paint and outdoor paint. It is also used as a precursor for cosmetics, colored glaze and dope. It finds application in the tin-lead plating. It is also useful as a stabilizing agent for vinyl chloride plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
MDL Number MFCD00078155

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% (Metal Basis)
UN Number 1479
Synonym White L ead, C.I. 77597
Solubility Information Slightly soluble in carbon dioxide solution,acetic and dilute nitric acid. Insoluble in water and ethanol.
Formula Weight 775.63
Appearance White powder
Density 6.14 g/cm3 (at 20°C)
Chemical Name or Material Lead(II) Carbonate, Basic
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1; Packaging Group: III
EINECSNumber 215-290-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only