missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Bromide [for Perovskite precursor], TCI America™

Catalog No. L02881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L02881G Supplier TCI America Supplier No. L02881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
SMILES [Br-].[Br-].[Pb++]

Specifications

Melting Point 373°C
Color White
Physical Form Crystalline Powder
Quantity 1 g
UN Number 2291
Formula Weight 367.01
Chemical Name or Material Lead(II) Bromide [for Perovskite precursor]