missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) bromide, 98+%

Catalog No. AAA1940622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1940622 100 g
AAA1940636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1940622 Supplier Thermo Scientific Chemicals Supplier No. A1940622
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 10031-22-8 | Br2Pb | 367.00 g/mol

It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

Solubility
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
SMILES [Br-].[Br-].[Pb++]

Specifications

Boiling Point 916°C
Melting Point 373°C
Quantity 100 g
Linear Formula PbBr2
UN Number UN2291
Merck Index 14,5403
Solubility Information Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.
IUPAC Name dibromolead
Formula Weight 367.01
Percent Purity ≥98%
Odor Odorless
Density 6.66
Chemical Name or Material Lead(II) bromide
Show More Show Less
Hazard Category H302+H332-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 233-084-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only