missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) acetate, basic
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Linear Formula | Pb(OOCCH3)2·2Pb(OH)2 |
| Quantity | 500 g |
| UN Number | UN1616 |
| Sensitivity | Air sensitive |
| Merck Index | 14,5419 |
| Formula Weight | 807.72 |
| Chemical Name or Material | Lead(II) acetate, basic |
Safety and Handling
| Hazard Category | H351-H360Df-H373-H500 |
| Hazard Statement | GHS H Statement H360-H351-H373 May damage fertility or the unborn child. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P260-P281-P308+P313-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE |
| EINECSNumber | 215-630-3 |
| RTECSNumber | OF8750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only