missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) 2,4-pentanedionate

Catalog No. p-7080528
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8985809 10 g
AA8985818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8985809 Supplier Thermo Scientific Chemicals Supplier No. 08985809
Only null left

CAS: 15282-88-9 | C10H14O4Pb | 405.418 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15282-88-9
Molecular Formula C10H14O4Pb
Molecular Weight (g/mol) 405.418
MDL Number MFCD00013499
InChI Key UNNUWSQNTAFLDC-SYWGCQIGSA-L
Synonym lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
PubChem CID 53393517
IUPAC Name bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
SMILES CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C

Specifications

Melting Point 141°C to 144°C
Odor Odorless
Quantity 10 g
UN Number UN3467
Sensitivity Hygroscopic
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 405.41
Physical Form Crystalline
Chemical Name or Material Lead (II) 2,4-pentanedionate
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
EINECSNumber 239-323-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only