missing translation for 'onlineSavingsMsg'
Learn More

Lead granules, 99.999% (metals basis)

Catalog No. AA4292818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4292818 50 g
AA4292830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4292818 Supplier Thermo Scientific Chemicals Supplier No. 04292818

CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Appearance Gray granules
Quantity 50 g
Odor Odorless
Form Granular
Assay Percent Range 99.999% (Metals basis)
Chemical Name or Material Lead
Hazard Category H350-H360FD-H362-H372
Precautionary Statement P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
EINECSNumber 231-100-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only