missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Lead-Free Density Blocks
For studying density and specific heat without using lead.
Supplier: Eisco™ PH0108PCBLF
Description
- Hook on top of cubes help to measure mass with spring scale
- Hooks allow to add and remove masses from boiling water for specific heat experiments
- Block of brass, copper, aluminum, steel included
- Instruction manual provided
- Contains three student activities with capture sheets and lab practical quiz
- Each mass is 32mm cubed
Specifications
| Density, Specific Gravity, Specific Heat Activities | |
| 32 mm | |
| Four cubes with hooks: aluminum, brass, copper, steel |
| Density Cube Set | |
| 1.26 in. |