missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Weight | ≈14.2g/25x25mm |
| Form | Foil |
| Percent Purity | 99.998% |
| Grade | Puratronic |
| Assay | (metals basis) |
| Chemical Name or Material | Lead foil, 2.0mm (0.08 in.) thick |
Safety and Handling
| Hazard Category | H350-H360FD-H362-H372 |
| Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c |
| EINECSNumber | 231-100-4 |
| RTECSNumber | OF7525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only