missing translation for 'onlineSavingsMsg'
Learn More

Lead foil, 0.1mm (0.004in) thick, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048916
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
Catalog No. Quantity
AA12051FI 50 x 50 mm
AA12051GH 100 x 100 mm
2 options

Catalog No. AA12051FI

Supplier: Thermo Scientific Chemicals 012051FI

Only null left

CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Quantity 50 x 50 mm
Odor Odorless
Weight ≈2.8g/50x50mm
Form Foil
Percent Purity 99.998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Lead foil, 0.1mm (0.004 in.) thick
Hazard Category H350-H360FD-H362-H372
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
EINECSNumber 231-100-4
RTECSNumber OF7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only