missing translation for 'onlineSavingsMsg'
Learn More

Lead Dioxide, Powder, 97%, Spectrum™ Chemical

Catalog No. 18611233
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611233 500 g Poly Bottle
18611231 25 g Amber Glass Bottle
18611228 100 g Poly Bottle
18611230 2.5 kg Poly Pail
18611229 12 kg Poly Pail
18611232 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611233 Supplier Spectrum Chemical Mfg Cor Supplier No. L1063500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 1309-60-0

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.