missing translation for 'onlineSavingsMsg'
Learn More

Lead Chromate, Powder, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L1055500GM

 View more versions of this product

Catalog No. 18611226


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7758-97-6
500 g
ACS
1
98%
Poly Bottle

Chemical Identifiers

7758-97-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.