missing translation for 'onlineSavingsMsg'
Learn More

Lead Chromate, Powder, ACS, 98%, Spectrum™ Chemical

Catalog No. 18611225
Change view
Click to view available options
Quantity:
125 g
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611225 125 g Poly Bottle
18611226 500 g Poly Bottle
18611227 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611225 Supplier Spectrum Chemical Mfg Cor Supplier No. L1055125GM

Chemical Identifiers

CAS 7758-97-6

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.