missing translation for 'onlineSavingsMsg'
Learn More

Lead Chloride, Powder, 97%, Spectrum™ Chemical

Catalog No. 18611218
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611218 500 g Poly Bottle
18611220 2.5 kg Poly Pail
18611217 12 kg Poly Pail
18611219 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611218 Supplier Spectrum Chemical Mfg Cor Supplier No. L1050500GM

Chemical Identifiers

CAS 7758-95-4

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.