missing translation for 'onlineSavingsMsg'
Learn More

Lead Antimony shot, 2mm (0.08in)

$93.04 - $93.04

Specifications

Chemical Name or Material Lead Antimony shot
Name Note 2mm (0.08 in.)
Odor Odorless
Form Shot
MDL Number MFCD00134050
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1022436
View Documents
Thermo Scientific Chemicals
01022436
500 g
Each for $93.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead Antimony shot
Odorless
MFCD00134050
2mm (0.08 in.)
Shot
UN3077
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only