missing translation for 'onlineSavingsMsg'
Learn More

Lead antimonide, 99+% (metals basis), Thermo Scientific™

$236.55 - $236.55

Chemical Identifiers

CAS 12266-38-5
Molecular Formula PbSb
Molecular Weight (g/mol) 328.96
MDL Number MFCD00151190
InChI Key QQHJESKHUUVSIC-UHFFFAOYSA-N
Synonym lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
PubChem CID 72720464
IUPAC Name stibanylidynelead
SMILES [Sb]#[Pb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3635006
View Documents
Thermo Scientific Chemicals
3635006
5 g
Each for $236.55
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12266-38-5
328.96
QQHJESKHUUVSIC-UHFFFAOYSA-N
72720464
[Sb]#[Pb]
PbSb
MFCD00151190
lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
stibanylidynelead

Specifications

12266-38-5
5 g
MFCD00151190
lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
[Sb]#[Pb]
328.96
328.94
Odorless
Lead antimonide
Lump
PbSb
UN1549
QQHJESKHUUVSIC-UHFFFAOYSA-N
stibanylidynelead
72720464
≥99%
6.72 g/mL
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

EINECSNumber : 235-536-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'