missing translation for 'onlineSavingsMsg'
Learn More

Lead, +99%, powder, Thermo Scientific Chemicals

Catalog No. AC198110010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
HDPE Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC198110010 1 kg HDPE Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC198110010 Supplier Thermo Scientific Chemicals Supplier No. 198110010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-92-1 | Molecular Formula: Pb | Molecular Weight: 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Melting Point 327.4°C
Boiling Point 1740.0°C
Quantity 1 kg
Packaging HDPE Plastic bottle
Form Powder
Assay Percent Range 99+%
Linear Formula Pb
Formula Weight 207.19
Percent Purity ≥99%
Chemical Name or Material Lead, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-100-4
RTECSNumber OF7525000
TSCA TSCA

RUO – Research Use Only