missing translation for 'onlineSavingsMsg'
Learn More

Lead, 99+%, granular

Catalog No. AC222625000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222625000 500 g Plastic bottle
AC222621000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222625000 Supplier Thermo Scientific Chemicals Supplier No. 222625000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Melting Point 327.4°C
Boiling Point 1740.0°C
Quantity 500 g
Packaging Plastic bottle
Form Granular
Assay Percent Range 99+%
Linear Formula Pb
Fieser 02,233
Merck Index 15, 5451
Formula Weight 207.19
Percent Purity ≥99%
Chemical Name or Material Lead, p.a.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May damage fertility. May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May cause harm to breast-fed children.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Avoid contact during pregnancy/while nursing.
EINECSNumber 231-100-4
RTECSNumber OF7525000
TSCA TSCA

RUO – Research Use Only