missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LDV
Description
LDV is a α4β1 integrin (VLA-4) ligand (Kd ~ 12 nM). Non-fluorescent derivative of LDV FITC (Cat. No. 4577).
Specifications
Specifications
| CAS | 1155866-55-9 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 979.13 |
| Quantity | 1 mg |