missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ LDPE Carboy

Catalog No. 14388176
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14388176 50 L
14388174 10 L
14388175 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14388176 Supplier Fisherbrand™ Supplier No. 14388176
Only null left

Store and transport reagents; Closure Material: Polypropylene; Color: White

  • Store and transport reagents with 10 L FISHERBRAND LDPE, Carboy, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands.
  • These carboys accept 13.5 rubber stoppers for an alternative sealing method.
  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof
  • Includes:White polypropylene screw cap and 83B closure

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 50 L
Color Natural
Disposable Reusable
Material LDPE
Mouth Style Wide
Product Type Carboy
Sterility Non-sterile