missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lck Inhibitor II

Catalog No. AAJ64111LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64111LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64111LB0 Supplier Thermo Scientific Chemicals Supplier No. J64111LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A cell-permeable, ATP binding site-targeting, tri-substituted pyrimidine that acts as a highly potent Lck (lymphocyte specific kinase) inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 918870-43-6
Molecular Formula C24H26N6O3
Synonym 3-(2-(1H-Benzo[d]imidazol-1-yl)-6-(2-morpholinoethoxy)-pyrimidin-4-ylamino)-4-methylphenol;

Specifications

Quantity 5 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, ATP binding site-targeting, tri-substituted pyrimidine that acts as a highly potent Lck (lymphocyte specific kinase) inhibitor
Formula Weight 446.50
Sensitivity Light Sensitive
Product Type Lck Inhibitor II
Form Solid

RUO – Research Use Only