Learn More
Enzo Life Sciences LC3B monoclonal antibody (5F10) (100 g) DFS Item
Supplier:  Enzo ALX803080C100
Clone: 5F10. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the N-terminus of LC3B conjugated to hemocyanin UniProt: Q9GZQ8. Species Reactivity: Human, Mouse, Rat, Hamster, Dog. Specificity: Recognizes both forms of endogenous LC3B. Applications: ICC, WB. Purity: Purified from serum-free cell culture supernatant by subsequent ultrafiltration and size exclusion chromatography. Formulation: Lyophilized from 1ml PBS containing PEG, sucrose and 0.09% sodium azide.
Catalog No. 501031758
Only null left
Explore available promotions