missing translation for 'onlineSavingsMsg'
Learn More

LC-MS Acetonitrile, OmniSolv, MilliporeSigma™

Catalog No. MAX01561
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX01561 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX01561 Supplier Technidata Supplier No. US1AX0156/1
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
SMILES CC#N

Specifications

Quantity 4 L
Physical Form Liquid
Assay Percent Range 99.9% min.
Chemical Name or Material LC-MS Acetonitrile, LC-MS OmniSolv™
Recommended Storage Room Temperature