Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Description
- Each measurement combines 27 images illuminated with different colours and from different directions
- Enables preview of measurement area and storage and recall of image after measurement
- Measures and stores up to 20 reference standards with each date and time stamped with option to name
- Stores up to 350 sample measurements on device and compares to reference standards
- Samples can be tagged via on-screen keyboard for text or via built in voice recorder
- For indoor use only
LC 100 and SV 100 Integrated Kit
- Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifications
Specifications
| Display | TFT |
| For Use With (Application) | Food, Bev, Cosmetics, Pharm |
| Includes | Spectrophotometer, holder, sample cells, USB cord, carrying case |
| Measurement Time | 1.8 sec. |