missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Broth, Miller (Pre-Buffered Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
$469.51 - $469.51
Specifications
| Format | Pre-Buffered Capsules |
|---|---|
| Media Type | LB Broth |
| Product Type | Microbiology Media |
| Formulation | Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, TRIS/TRIS HCl 1.5g/L |
| For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|
BP9731-500
|
Fisher BioReagents™
BP9731500 |
25 x 25 g | Poly Bottle |
Each for $469.51
|
|
||||
Description
- Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 1g of broth
- Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
- pH will vary with temperature
Specifications
| Pre-Buffered Capsules | |
| Microbiology Media | |
| For maintenance and propagation of Escherichia coli in molecular biology procedures |
| LB Broth | |
| Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, TRIS/TRIS HCl 1.5g/L |