missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth, Miller (Granulated)

Catalog No. bp97235
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 kg
500 g
Packaging Type:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging Type
BP97235 5 kg Poly Pail
BP9723500 500 g Poly Bottle
2 options

Catalog No. BP97235

Supplier: Fisher BioReagents™ BP9723 5

Only null left

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Format: Granulated powder

Specifications

Form Granulated Powder
Media Type LB Broth
Product Type Bacterial Culture
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Pail
Quantity 5 kg