missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Broth, Miller (Granulated)
Description
- Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
- Format: Granulated powder
Specifications
Specifications
| Form | Granulated Powder |
| Media Type | LB Broth |
| Product Type | Microbiology Media |
| Formulation | Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L |
| For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |