missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LB Broth (1X)
GREENER_CHOICE

Catalog No. 50591332
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50591332 1 L
10855021 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50591332 Supplier Gibco™ Supplier No. 10855001

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture.

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized and can be used for nearly all applications that require a liquid bacteria culturing step, such as protein expression, cloning, and expansion.

LB broth is one of the most referenced bacterial media for cultivating and maintaining E. coli strains. Our LB broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium.

LB broth formulation

Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.

  • Tryptone provides nitrogen, vitamins, minerals, and amino acids essential for growth
  • Yeast extract is a source of vitamins, particularly B-group vitamins
  • Sodium chloride supplies essential electrolytes for transport and osmotic balance

Primary applications

  • Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
  • Resuspension of T. pyriformis cell fractions

Gibco media bottles

Each media bottle features an angled opening for simplified pipetting. The 1-L Gibco media bottle uses 39% less source material (33% less for the 500-mL bottle) than comparable media bottles on the market. The cap and bottle are made from high-density polyethylene (HDPE) and polyethylene terephthalate (PETE) and can both be recycled.

Find the media format that you need

We offer other ready-to-use media formulations, such as Terrific Broth (Cat. No. A1374301) or S.O.C Recovery Medium (Cat. No. 15544034).

Specifications

Content And Storage • 1 L LB Broth (1X)

Store at room temperature.

Format Bottle
Media Type LB Broth (Lennox)
Preparation Method Ready-to-Use
Product Type Liquid Medium
Sterility Sterile
Target Organism Class Coliforms
Quantity 1 L
Shipping Condition Room Temperature
Species Escherichia coli
What additional materials do I need for my transformation?

You will need bacterial growth media, such as LB, the bacterial selection antibiotic, and induction reagent (IPTG, X-gal). We offer imMedia growth medium which is a pre-mixed, pre-sterilized medium containing selection antibiotics for growth of E. coli strains. It can be prepared with or without IPTG and X-gal.

What bacterial growth media should I use?

Check out our bacterial growth media chart (https://www.thermofisher.com/us/en/home/life-science/cell-culture/microbiological-culture/bacterial-growth-media.html).

What is the optimal pH of low salt LB for LB + blasticidin plates?

We recommend a pH of 7 or less and half the normal amount of NaCl in your LB media or plates.

See the following paper for details on optimal conditions: Yamaguchi et al (1965) Inhibition of Protein Synthesis by Blasticidin S. Journal of Biochemistry (Tokyo) Volume 57: pp 667-677.

For Research Use Only. Not for use in diagnostic procedures.