missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals LB Agar plate with 50μg/mL Carbenicillin
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used for growth of Escherichia coli, plasmid and other bacterial cultures
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Media Type | LB Agar |
| For Use With (Application) | For growth of Escherichia coli, plasmid and other bacterial cultures |
| Packaging Type | Plate |
| Quantity | 20 Plates |
| Height (Metric) | 15 mm |
| Diameter (Metric) Outside | 10 cm |
RUO – Research Use Only