missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals LB Agar plate with 10μg/mL Tetracycline

Catalog No. AAJ62894EQF
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62894EQF 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62894EQF Supplier Thermo Scientific Chemicals Supplier No. J62894EQF

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Used for growth of Escherichia coli, plasmid and other bacterial cultures

Specifications

Content And Storage 2°C to 8°C
Form Solid
Media Type LB Agar
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging Type Plate
Quantity 20 Plates
Height (Metric) 15 mm
Diameter (Metric) Outside 10 cm

RUO – Research Use Only